Assay Details
| Gene Symbol: AT4G14805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148322.2 | NP_680688.1 | 2 - 3 | 434 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G14805| Gene Symbol: AT4G14805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148322.2 | NP_680688.1 | 2 - 3 | 434 | 72 | |
Set Membership: |
|