Assay Details
| Gene Symbol: XIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119015.4 | NP_194600.2 | 32 - 33 | 4274 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XIH| Gene Symbol: XIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119015.4 | NP_194600.2 | 32 - 33 | 4274 | 143 | |
Set Membership: |
|