Assay Details
| Gene Symbol: AT4G35165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148398.3 | NP_680764.1 | 1 | 319 | 82 | |
| GenBank mRNA | DQ132713.1 | - | 1 | 319 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G35165| Gene Symbol: AT4G35165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148398.3 | NP_680764.1 | 1 | 319 | 82 | |
| GenBank mRNA | DQ132713.1 | - | 1 | 319 | 82 | |
Set Membership: |
|