Assay Details
| Gene Symbol: TRFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148439.4 | NP_683280.1 | 2 - 3 | 978 | 76 | |
| GenBank mRNA | AY519534.1 | - | 2 - 3 | 870 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRFL7| Gene Symbol: TRFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148439.4 | NP_683280.1 | 2 - 3 | 978 | 76 | |
| GenBank mRNA | AY519534.1 | - | 2 - 3 | 870 | 76 | |
Set Membership: |
|