Assay Details
| Gene Symbol: BGAL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332978.1 | NP_001319124.1 | 5 - 6 | 661 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGAL15| Gene Symbol: BGAL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332978.1 | NP_001319124.1 | 5 - 6 | 661 | 127 | |
Set Membership: |
|