Assay Details
| Gene Symbol: CAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102709.4 | NP_564333.1 | 2 - 3 | 814 | 83 | |
| GenBank mRNA | AY074857.1 | - | 2 - 3 | 785 | 83 | |
| BT030337.1 | - | 2 - 3 | 522 | 83 | ||
More Information
Set Membership: |
|

CAD1
Genomic Map