Assay Details
| Gene Symbol: AT1G29770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102717.4 | NP_174270.1 | 1 | 978 | 89 | |
| GenBank mRNA | BT012604.1 | - | 1 | 797 | 89 | |
| BT014842.1 | - | 1 | 635 | 89 | ||
More Information
Set Membership: |
|

AT1G29770
Genomic Map