Assay Details
| Gene Symbol: AT1G29790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102719.7 | NP_174272.2 | 1 - 2 | 1462 | 151 | |
| GenBank mRNA | AY093204.1 | - | 1 - 2 | 1204 | 151 | |
More Information
Set Membership: |
|

AT1G29790
Genomic Map