Assay Details
| Gene Symbol: RPL18AA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084157.1 | NP_001077626.1 | 3 - 4 | 422 | 93 | |
| NM_102737.3 | NP_564343.1 | 3 - 4 | 493 | 93 | ||
| GenBank mRNA | AK229311.1 | - | 3 - 4 | 448 | 93 | |
| BT030607.1 | - | 3 - 4 | 306 | 93 | ||
More Information
Set Membership: |
|

RPL18AA
Genomic Map