Assay Details
| Gene Symbol: AT1G30260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102765.4 | NP_564352.1 | 1 | 754 | 74 | |
| GenBank mRNA | AY057696.1 | - | 1 | 566 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G30260| Gene Symbol: AT1G30260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102765.4 | NP_564352.1 | 1 | 754 | 74 | |
| GenBank mRNA | AY057696.1 | - | 1 | 566 | 74 | |
Set Membership: |
|