Assay Details
| Gene Symbol: CIPK23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332883.1 | NP_001320726.1 | 13 - 14 | 1731 | 140 | |
| NM_001332884.1 | NP_001320727.1 | 13 - 14 | 1867 | 140 | ||
| NM_102766.6 | NP_564353.1 | 13 - 14 | 1785 | 140 | ||
| GenBank mRNA | AY035226.1 | - | 13 - 14 | 1315 | 140 | |
| AY056419.1 | - | 13 - 14 | 1687 | 140 | ||
| AY090322.1 | - | 13 - 14 | 1315 | 140 | ||
More Information
Set Membership: |
|

CIPK23
Genomic Map