Assay Details
| Gene Symbol: CIPK23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332883.1 | NP_001320726.1 | 2 - 3 | 702 | 84 | |
| NM_102766.6 | NP_564353.1 | 2 - 3 | 756 | 84 | ||
| GenBank mRNA | AY035226.1 | - | 2 - 3 | 286 | 84 | |
| AY056419.1 | - | 2 - 3 | 658 | 84 | ||
| AY090322.1 | - | 2 - 3 | 286 | 84 | ||
More Information
Set Membership: |
|

CIPK23
Genomic Map