Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036038.2 | NP_001031115.1 | 7 - 8 | 1274 | 77 | |
| NM_001332891.1 | NP_001320924.1 | 6 - 7 | 1510 | 77 | ||
| NM_102771.4 | NP_174323.1 | 6 - 7 | 1379 | 77 | ||
| GenBank mRNA | AF013467.1 | - | 6 - 7 | 1304 | 77 | |
| AY669788.1 | - | 6 - 7 | 558 | 77 | ||
More Information
Set Membership: |
|

ARF6
Genomic Map