Assay Details
| Gene Symbol: AT1G30750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102811.2 | NP_564361.1 | 1 | 659 | 91 | |
| GenBank mRNA | AY050922.1 | - | 1 | 646 | 91 | |
| AY113970.1 | - | 1 | 623 | 91 | ||
More Information
Set Membership: |
|

AT1G30750
Genomic Map