Assay Details
| Gene Symbol: AT1G30820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102819.4 | NP_174368.1 | 19 - 20 | 1810 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G30820| Gene Symbol: AT1G30820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102819.4 | NP_174368.1 | 19 - 20 | 1810 | 143 | |
Set Membership: |
|