Assay Details
| Gene Symbol: DIS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102820.3 | NP_564364.1 | 5 - 6 | 769 | 92 | |
| GenBank mRNA | AK118475.1 | - | 5 - 6 | 733 | 92 | |
| AY084256.1 | - | 5 - 6 | 733 | 92 | ||
| BT005308.1 | - | 5 - 6 | 571 | 92 | ||
More Information
Set Membership: |
|

DIS2
Genomic Map