Assay Details
Gene Symbol: AK-HSDH I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102861.3 | NP_174408.1 | 15 - 16 | 2432 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
