Assay Details
| Gene Symbol: RECQL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084170.2 | NP_001077639.1 | 17 - 18 | 1686 | 96 | |
| NM_001332956.1 | NP_001319121.1 | 17 - 18 | 1733 | 96 | ||
| NM_001332959.1 | NP_001322843.1 | 17 - 18 | 1733 | 96 | ||
| NM_102875.4 | NP_174421.3 | 17 - 18 | 1737 | 96 | ||
| GenBank mRNA | AJ404471.1 | - | 17 - 18 | 1722 | 96 | |
More Information
Set Membership: |
|

RECQL2
Genomic Map