Assay Details
| Gene Symbol: RECQL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084170.2 | NP_001077639.1 | 6 - 7 | 914 | 66 | |
| NM_001332956.1 | NP_001319121.1 | 6 - 7 | 961 | 66 | ||
| NM_001332957.1 | NP_001322841.1 | 6 - 7 | 961 | 66 | ||
| NM_001332958.1 | NP_001322840.1 | 6 - 7 | 961 | 66 | ||
| NM_001332959.1 | NP_001322843.1 | 6 - 7 | 961 | 66 | ||
| NM_001332960.1 | NP_001322842.1 | 6 - 7 | 961 | 66 | ||
| NM_001332961.1 | NP_001322844.1 | 6 - 7 | 960 | 66 | ||
| NM_102875.4 | NP_174421.3 | 6 - 7 | 965 | 66 | ||
| GenBank mRNA | AJ404471.1 | - | 6 - 7 | 950 | 66 | |
More Information
Set Membership: |
|

RECQL2
Genomic Map