Assay Details
Gene Symbol: ROPGEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332971.1 | NP_001320846.1 | 1 - 2 | 727 | 78 | |
NM_102900.5 | NP_174446.2 | 3 - 4 | 928 | 78 | ||
GenBank mRNA | AK222010.1 | - | 3 - 4 | 875 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
