Assay Details
| Gene Symbol: AT1G31660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102901.4 | NP_174447.2 | 4 - 5 | 621 | 125 | |
| GenBank mRNA | AY091144.1 | - | 4 - 5 | 620 | 125 | |
| BT003812.1 | - | 4 - 5 | 521 | 125 | ||
More Information
Set Membership: |
|

AT1G31660
Genomic Map