Assay Details
| Gene Symbol: XLG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198196.1 | NP_001185125.1 | 4 - 5 | 1462 | 64 | |
| NM_001332988.1 | NP_001319126.1 | 4 - 5 | 1574 | 64 | ||
| NM_001332989.1 | NP_001320661.1 | 4 - 5 | 1664 | 64 | ||
| NM_001332990.1 | NP_001320662.1 | 3 - 4 | 1698 | 64 | ||
| NM_102929.2 | NP_174475.1 | 5 - 6 | 1484 | 64 | ||
| NM_179406.2 | NP_849737.1 | 4 - 5 | 1537 | 64 | ||
| GenBank mRNA | AK229561.1 | - | 4 - 5 | 1457 | 64 | |
| AK317243.1 | - | 4 - 5 | 1308 | 64 | ||
| AK317663.1 | - | 3 - 4 | 1227 | 64 | ||
| GQ229390.1 | - | 5 - 6 | 1487 | 64 | ||
More Information
Set Membership: |
|

XLG3
Genomic Map