Assay Details
| Gene Symbol: SCAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102939.4 | NP_174485.1 | 8 - 9 | 813 | 72 | |
| GenBank mRNA | AK226594.1 | - | 1544 | 72 | ||
| AY087388.1 | - | 8 - 9 | 717 | 72 | ||
| BT025793.1 | - | 8 - 9 | 582 | 72 | ||
More Information
Set Membership: |
|

SCAMP5
Genomic Map