Assay Details
| Gene Symbol: NHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102967.3 | NP_174512.1 | 4 - 5 | 1624 | 70 | |
| GenBank mRNA | AY099548.1 | - | 4 - 5 | 1563 | 70 | |
| BT002115.1 | - | 4 - 5 | 1541 | 70 | ||
More Information
Set Membership: |
|

NHL8
Genomic Map