Assay Details
| Gene Symbol: STY46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342498.1 | NP_001328294.1 | 5 - 6 | 933 | 81 | |
| NM_001342499.1 | NP_001328293.1 | 5 - 6 | 933 | 81 | ||
| NM_120008.2 | NP_568041.1 | 5 - 6 | 1119 | 81 | ||
More Information
Set Membership: |
|

STY46
Genomic Map