Assay Details
| Gene Symbol: BGAL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160820.1 | NP_001154292.1 | 5 - 6 | 816 | 70 | |
| NM_120020.4 | NP_195571.3 | 5 - 6 | 868 | 70 | ||
More Information
Set Membership: |
|

BGAL14
Genomic Map