Assay Details
| Gene Symbol: BGAL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160820.1 | NP_001154292.1 | 6 - 7 | 922 | 62 | |
| NM_120020.4 | NP_195571.3 | 6 - 7 | 974 | 62 | ||
More Information
Set Membership: |
|

BGAL14
Genomic Map