Assay Details
| Gene Symbol: KAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342508.1 | NP_001329354.1 | 5 - 6 | 1864 | 68 | |
| NM_120021.2 | NP_195572.2 | 5 - 6 | 1416 | 68 | ||
| NM_179236.3 | NP_849567.2 | 5 - 6 | 1864 | 68 | ||
| GenBank mRNA | AY265959.1 | - | 5 - 6 | 1456 | 68 | |
More Information
Set Membership: |
|

KAK
Genomic Map