Assay Details
| Gene Symbol: RPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120024.3 | NP_195575.1 | 2 - 3 | 270 | 77 | |
| GenBank mRNA | AF360319.1 | - | 2 - 3 | 212 | 77 | |
| AY113889.1 | - | 1 - 2 | 131 | 77 | ||
| U33269.1 | - | 2 - 3 | 151 | 77 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

RPN10
Genomic Map