Assay Details
| Gene Symbol: RPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120024.3 | NP_195575.1 | 4 - 5 | 486 | 79 | |
| GenBank mRNA | AF360319.1 | - | 4 - 5 | 428 | 79 | |
| AY113889.1 | - | 3 - 4 | 347 | 79 | ||
| U33269.1 | - | 4 - 5 | 367 | 79 | ||
More Information
Set Membership: |
|

RPN10
Genomic Map