Assay Details
| Gene Symbol: VHA-A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120068.5 | NP_568051.1 | 14 - 15 | 2030 | 68 | |
| GenBank mRNA | AK227321.1 | - | 14 - 15 | 1997 | 68 | |
| AY060557.1 | - | 14 - 15 | 1991 | 68 | ||
| BT002615.1 | - | 14 - 15 | 1916 | 68 | ||
More Information
Set Membership: |
|

VHA-A3
Genomic Map