Assay Details
| Gene Symbol: VHA-A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120068.5 | NP_568051.1 | 2 - 3 | 312 | 62 | |
| GenBank mRNA | AK227321.1 | - | 2 - 3 | 279 | 62 | |
| AY060557.1 | - | 2 - 3 | 273 | 62 | ||
| BT002615.1 | - | 2 - 3 | 198 | 62 | ||
More Information
Set Membership: |
|

VHA-A3
Genomic Map