Assay Details
| Gene Symbol: VHA-A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120068.5 | NP_568051.1 | 4 - 5 | 541 | 98 | |
| GenBank mRNA | AK227321.1 | - | 4 - 5 | 508 | 98 | |
| AY060557.1 | - | 4 - 5 | 502 | 98 | ||
| BT002615.1 | - | 4 - 5 | 427 | 98 | ||
More Information
Set Membership: |
|

VHA-A3
Genomic Map