Assay Details
| Gene Symbol: IMPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342523.1 | NP_001320164.1 | 6 - 7 | 769 | 90 | |
| NM_120072.5 | NP_195623.3 | 6 - 7 | 856 | 90 | ||
| GenBank mRNA | AK175533.1 | - | 6 - 7 | 797 | 90 | |
| AK220687.1 | - | 6 - 7 | 797 | 90 | ||
| BT010891.1 | - | 6 - 7 | 769 | 90 | ||
More Information
Set Membership: |
|

IMPL2
Genomic Map