Assay Details
| Gene Symbol: APL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342527.1 | NP_001328715.1 | 10 - 11 | 1145 | 121 | |
| NM_120081.2 | NP_195632.1 | 10 - 11 | 1323 | 121 | ||
| GenBank mRNA | AY059862.1 | - | 10 - 11 | 1263 | 121 | |
| BT010378.1 | - | 9 - 10 | 1104 | 121 | ||
More Information
Set Membership: |
|

APL3
Genomic Map