Assay Details
| Gene Symbol: APL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342527.1 | NP_001328715.1 | 14 - 15 | 1508 | 85 | |
| NM_120081.2 | NP_195632.1 | 14 - 15 | 1686 | 85 | ||
| GenBank mRNA | AY059862.1 | - | 14 - 15 | 1626 | 85 | |
| BT010378.1 | - | 13 - 14 | 1467 | 85 | ||
More Information
Set Membership: |
|

APL3
Genomic Map