Assay Details
| Gene Symbol: APL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342527.1 | NP_001328715.1 | 2 - 3 | 257 | 74 | |
| NM_120081.2 | NP_195632.1 | 2 - 3 | 435 | 74 | ||
| GenBank mRNA | AY059862.1 | - | 2 - 3 | 375 | 74 | |
| BT010378.1 | - | 1 - 2 | 216 | 74 | ||
More Information
Set Membership: |
|

APL3
Genomic Map