Assay Details
| Gene Symbol: APL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342527.1 | NP_001328715.1 | 6 - 7 | 726 | 132 | |
| NM_120081.2 | NP_195632.1 | 6 - 7 | 904 | 132 | ||
| GenBank mRNA | AY059862.1 | - | 6 - 7 | 844 | 132 | |
| BT010378.1 | - | 5 - 6 | 685 | 132 | ||
More Information
Set Membership: |
|

APL3
Genomic Map