Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120087.4 | NP_195637.1 | 1 - 2 | 360 | 69 | |
| NM_179192.2 | NP_849523.1 | 1 - 2 | 223 | 69 | ||
| NM_179193.2 | NP_849524.1 | 1 - 2 | 166 | 69 | ||
| NM_179194.1 | NP_849525.1 | 1 - 2 | 161 | 69 | ||
| GenBank mRNA | AK316798.1 | - | 1 - 2 | 164 | 69 | |
| L00649.1 | - | 1 - 2 | 135 | 69 | ||
| Z14988.1 | - | 135 | 69 | |||
More Information
Set Membership: |
|

CCR1
Genomic Map