Assay Details
| Gene Symbol: CAD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036735.2 | NP_001031812.1 | 3 - 4 | 776 | 106 | |
| NM_120093.5 | NP_195643.1 | 3 - 4 | 776 | 106 | ||
| GenBank mRNA | AF370498.1 | - | 3 - 4 | 772 | 106 | |
| AY064669.1 | - | 3 - 4 | 726 | 106 | ||
| AY302076.1 | - | 3 - 4 | 726 | 106 | ||
More Information
Set Membership: |
|

CAD9
Genomic Map