Assay Details
| Gene Symbol: CAD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120093.5 | NP_195643.1 | 4 - 5 | 930 | 64 | |
| GenBank mRNA | AF370498.1 | - | 4 - 5 | 926 | 64 | |
| AY064669.1 | - | 4 - 5 | 880 | 64 | ||
| AY302076.1 | - | 4 - 5 | 880 | 64 | ||
More Information
Set Membership: |
|

CAD9
Genomic Map