Assay Details
| Gene Symbol: SRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127017.2 | NP_179060.1 | 1 - 2 | 493 | 124 | |
| GenBank mRNA | AY072119.1 | - | 1 - 2 | 494 | 124 | |
| AY122942.1 | - | 1 - 2 | 477 | 124 | ||
More Information
Set Membership: |
|

SRP2
Genomic Map