Assay Details
| Gene Symbol: LIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127084.3 | NP_179126.2 | 4 - 5 | 684 | 113 | |
| GenBank mRNA | AF530477.1 | - | 4 - 5 | 525 | 113 | |
| AK229578.1 | - | 4 - 5 | 583 | 113 | ||
| BT025547.1 | - | 4 - 5 | 498 | 113 | ||
More Information
Set Membership: |
|

LIP1
Genomic Map