Assay Details
| Gene Symbol: AT2G15240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127085.5 | NP_565370.1 | 1 - 2 | 307 | 108 | |
| GenBank mRNA | AF378872.1 | - | 1 - 2 | 174 | 108 | |
| AY052736.1 | - | 1 - 2 | 95 | 108 | ||
More Information
Set Membership: |
|

AT2G15240
Genomic Map