Assay Details
| Gene Symbol: AT2G15240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127085.5 | NP_565370.1 | 4 - 5 | 479 | 67 | |
| GenBank mRNA | AF378872.1 | - | 4 - 5 | 346 | 67 | |
| AY052736.1 | - | 4 - 5 | 267 | 67 | ||
More Information
Set Membership: |
|

AT2G15240
Genomic Map