Assay Details
| Gene Symbol: MBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124852.5 | NP_001118324.1 | 5 - 6 | 2448 | 85 | |
| NM_001202601.1 | NP_001189530.1 | 5 - 6 | 2026 | 85 | ||
| NM_001335453.1 | NP_001325050.1 | 4 - 5 | 2310 | 85 | ||
| NM_001335454.1 | NP_001325052.1 | 5 - 6 | 2021 | 85 | ||
| NM_001335455.1 | NP_001325051.1 | 2 - 3 | 1833 | 85 | ||
| NM_127113.4 | NP_179155.1 | 4 - 5 | 2087 | 85 | ||
| NM_201741.2 | NP_973470.1 | 4 - 5 | 2291 | 85 | ||
More Information
Set Membership: |
|

MBR1
Genomic Map