Assay Details
| Gene Symbol: MBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124852.5 | NP_001118324.1 | 7 - 8 | 2700 | 103 | |
| NM_001202601.1 | NP_001189530.1 | 7 - 8 | 2278 | 103 | ||
| NM_001335453.1 | NP_001325050.1 | 6 - 7 | 2562 | 103 | ||
| NM_001335454.1 | NP_001325052.1 | 7 - 8 | 2273 | 103 | ||
| NM_001335455.1 | NP_001325051.1 | 4 - 5 | 2085 | 103 | ||
| NM_127113.4 | NP_179155.1 | 6 - 7 | 2339 | 103 | ||
| NM_201741.2 | NP_973470.1 | 6 - 7 | 2543 | 103 | ||
More Information
Set Membership: |
|

MBR1
Genomic Map