Assay Details
| Gene Symbol: AT2G15710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127133.1 | NP_179173.1 | 5 - 6 | 838 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G15710| Gene Symbol: AT2G15710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127133.1 | NP_179173.1 | 5 - 6 | 838 | 142 | |
Set Membership: |
|