Assay Details
| Gene Symbol: DRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335476.1 | NP_001324537.1 | 2 - 3 | 711 | 121 | |
| NM_127193.2 | NP_179232.1 | 1 - 2 | 937 | 121 | ||
More Information
Set Membership: |
|

DRD1
Genomic Map