Assay Details
| Gene Symbol: NERD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335477.1 | NP_001324854.1 | 3 - 4 | 2319 | 83 | |
| NM_127202.5 | NP_179241.4 | 3 - 4 | 2240 | 83 | ||
More Information
Set Membership: |
|

NERD
Genomic Map